Learn More
Hach Company Cylinder, Graduated, Double Metric, 100 mL
Supplier:  Hach Company 2928302
Encompass
These glass graduated cylinders feature two sets of graduations for reading either up or down Calibrated "to deliver"
Catalog No. 50-199-6811