Learn More
Chemglass Life Sciences Cylinder, 100mL, Red Stripe, Reinforcement Bead
SDP
Catalog No. NC1360309
Encompass_Preferred
Cylinder, 100mL, Red Stripe, Reinforcement Bead. Calibrated “To Contain”. A reinforced bead of glass near the top helps to reduce breakage, if the cylinder upsets. The 100mL cylinder is in accordance with ASTM E-133 and E-1272 and is used in ASTM D-86, D-216, and D-447.
Catalog No. NC1360309 Supplier Chemglass Life Sciences Supplier No. CG8254100
Cylinder, 100mL, Red Stripe, Reinforcement Bead. Calibrated “To Contain”. A reinforced bead of glass near the top helps to reduce breakage, if the cylinder upsets. The 100mL cylinder is in accordance with ASTM E-133 and E-1272 and is used in ASTM D-86, D-216, and D-447.