Learn More
Wilmad Labglass CYLINDER SOIL SUSPENSN 1000ML
Catalog No. 50804293
Encompass_Preferred
Cylinder, Soil Suspension, To Contain. Capacity: 1000mL; Approximate Height: 460mm. Used in determining the amount of soil in dispersed suspensions as described in ADTM Method D 422 and AASHO Method T 88
Catalog No. 50-804-293 Supplier Wilmad Labglass Supplier No. LG5519100
Cylinder, Soil Suspension, To Contain. Capacity: 1000mL; Approximate Height: 460mm. Used in determining the amount of soil in dispersed suspensions as described in ADTM Method D 422 and AASHO Method T 88