Learn More
Anton Paar Cylinder system with Peltier temperaturecontrol, counter-cooling with air, no circulatorrequired, temperature range 0 ºC to +180 ºC.
Catalog No. 5000580445
Encompass_Preferred
Some advantages of a Peltier temperature device compared to a circulation thermostat are:• Faster heating and cooling rates• No extra space requirement for the thermostat• No thermal transfer liquids required• Maintenance free• Lower noise level• Less energy consumptionTechnical Data Unit 108642 C-PTD 180/AIR/QCMin. temperature °C 0Max. temperature °C 180Mean heating rate K/min 8Mean cooling rate K/min 4
Catalog No. 50-005-80445 Supplier Anton Paar Supplier No. 108642
Some advantages of a Peltier temperature device compared to a circulation thermostat are:• Faster heating and cooling rates• No extra space requirement for the thermostat• No thermal transfer liquids required• Maintenance free• Lower noise level• Less energy consumptionTechnical Data Unit 108642 C-PTD 180/AIR/QCMin. temperature °C 0Max. temperature °C 180Mean heating rate K/min 8Mean cooling rate K/min 4