Learn More
Brady™ Cylinder Tank Lockout
Catalog No. 19103974 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
19-103-974 Red
1 options
Catalog No. 19-103-974 Supplier Brady™ Supplier No. SD02M
Add to Cart
Add to Cart
Lockout cylinder tanks, including propane tanks on fork trucks and stand alone propane tanks. The Brady™ Cylinder Tank Lockout's special design allows it to be used in tight spaces.
  • Revolutionary new rugged polystrene plastic device is used to lockout cylinder tanks, including propane tanks on fork trucks and stand alone propane tanks
  • Special design allows device to be used in tight spaces
  • Valve stem hole is 1.25 in. in diameter
Specifications
Product Type Unthreaded Cylinder Tank
Color Red
Material Polystyrene
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.