Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 1-1/4" surface, 3/8" height, Aluminum
SDP
Catalog No. 501901497
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1-1/4"
Height: 3/8"
Catalog No. 50-190-1497 Supplier Electron Microscopy Sciences Supplier No. 7583090
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1-1/4"
Height: 3/8"