Learn More
Grainger Sandvik Coromant Cylindrical Collet, 20 mm Shank, 5 mm Clamping, 1 ea
Catalog No. 502758333
Encompass_Preferred
This cylindrical collet is designed for precision clamping in machining applications. It is compatible with hydraulic membrane chucks and features an axial coolant delivery system to support efficient operation.
  • Designed for use with hydraulic membrane chucks
  • Supports axial concentric coolant entry and output
  • Compatible with 20 mm machine connection diameter
  • Accommodates a 5 mm workpiece diameter
  • Built for high precision clamping requirements
Catalog No. 50-275-8333 Supplier Grainger Supplier No. 14Z188
This cylindrical collet is designed for precision clamping in machining applications. It is compatible with hydraulic membrane chucks and features an axial coolant delivery system to support efficient operation.
  • Designed for use with hydraulic membrane chucks
  • Supports axial concentric coolant entry and output
  • Compatible with 20 mm machine connection diameter
  • Accommodates a 5 mm workpiece diameter
  • Built for high precision clamping requirements