Learn More
Azzota Corp Cylindrical cuvette 500mm 14ml
Catalog No. 5000886967
Encompass_Preferred
CYLINDRICAL CUVETTE Pathlength - 500mm Inside width - 19mm Volume - 14 0ml Dimensions - 22x52 5mm Includes two screw cap stoppers Type G - Optical Glass (340nm-2500nm)
Catalog No. 50-008-86967 Supplier Azzota Corp Supplier No. G87SC
CYLINDRICAL CUVETTE Pathlength - 500mm Inside width - 19mm Volume - 14 0ml Dimensions - 22x52 5mm Includes two screw cap stoppers Type G - Optical Glass (340nm-2500nm)