Learn More
Enzo Life Sciences Cylindrospermopsin (100µg). CAS: 143545-90-8
SDP
Catalog No. 502008761
Encompass_Preferred
Tricyclic alkaloid hepatotoxin. Exhibits a completely different mechanism of toxicity than microcystins. Protein synthesis inhibitor. Might be carcinogenic. Inhibits pyrimidine nucleotide synthesis. Purity: ≥95% (HPLC). Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8761 Supplier Enzo Life Sciences Supplier No. ALX350149C100
Tricyclic alkaloid hepatotoxin. Exhibits a completely different mechanism of toxicity than microcystins. Protein synthesis inhibitor. Might be carcinogenic. Inhibits pyrimidine nucleotide synthesis. Purity: ≥95% (HPLC). Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C.