Learn More
Abraxis Cylindrospermopsin Dipstick Test, 20 tests, 0-10 ppb detection range, Rapid Immunochromatographic Assay, Fresh Water sample, Gold colloid antibody
SDP
Catalog No. NC1394966
Encompass
Cylindrospermopsin Dipstick Test, 20 tests, 0-10 ppb detection range, Rapid Immunochromatographic Assay, Fresh Water sample, Gold colloid antibody
Catalog No. NC1394966 Supplier Abraxis Supplier No. 520030
Cylindrospermopsin Dipstick Test, 20 tests, 0-10 ppb detection range, Rapid Immunochromatographic Assay, Fresh Water sample, Gold colloid antibody