Learn More
Industrial Scientific Corporation CYLNDR CALIBRTN GAS 25 PPM HS
Catalog No. NC3091613
Encompass
CYLINDER, CALIBRATION GAS, 25 PPM HYDROGEN SULFIDE, 100 PPM CARBON MONOXIDE, 35% PENTANE (25% LEL), 18% OYYGEN, ALUMINUM, 58L
Catalog No. NC3091613 Supplier Industrial Scientific Corporation Supplier No. 18109155
CYLINDER, CALIBRATION GAS, 25 PPM HYDROGEN SULFIDE, 100 PPM CARBON MONOXIDE, 35% PENTANE (25% LEL), 18% OYYGEN, ALUMINUM, 58L