Learn More
Chemglass Life Sciences Cylinder, Graduated, 100mL, 24/40 Outer Joint, "To Deliver"
SDP
Catalog No. CG122302
Encompass_Preferred
Cylinder, Graduated, 100mL, 24/40 Outer Joint. Cylinders are calibrated to contain at 20¿ C. Supplied without stopper. For standard taper stoppers see CG-3000. Tolerances and subdivisions are the same as CG-8242.
Catalog No. CG122302 Supplier Chemglass Life Sciences Supplier No. CG122302
Cylinder, Graduated, 100mL, 24/40 Outer Joint. Cylinders are calibrated to contain at 20¿ C. Supplied without stopper. For standard taper stoppers see CG-3000. Tolerances and subdivisions are the same as CG-8242.