Learn More
MSC Clarke Floor Scrubber, 28 in Cleaning Path, AGM Battery
Catalog No. 502836906
Encompass
This floor scrubber is a robust solution for efficient floor cleaning, offering a 28 in cleaning path and 23 gal solution and recovery tank capacities. Designed for walk-behind operation, it is self-propelled and utilizes battery power, making it versatile for various commercial and industrial settings.
  • High capacity and long run times for extended operation.
  • Simple operation with ergonomically placed controls.
  • Quiet operation, ideal for noise-sensitive environments.
  • Optional chemical mixing system ensures precise chemical and water mixture.
  • Pulse-modulated solution control delivers precise and consistent solutions.
  • Gimble-mounted brushes and pad drivers for even scrubbing on uneven floors.
  • Onboard battery charger for convenient recharging.
  • Green-certified equipment.
Catalog No. 50-283-6906 Supplier MSC Supplier No. 35706720
Add to Cart
Add to Cart
Request a Quote
This floor scrubber is a robust solution for efficient floor cleaning, offering a 28 in cleaning path and 23 gal solution and recovery tank capacities. Designed for walk-behind operation, it is self-propelled and utilizes battery power, making it versatile for various commercial and industrial settings.
  • High capacity and long run times for extended operation.
  • Simple operation with ergonomically placed controls.
  • Quiet operation, ideal for noise-sensitive environments.
  • Optional chemical mixing system ensures precise chemical and water mixture.
  • Pulse-modulated solution control delivers precise and consistent solutions.
  • Gimble-mounted brushes and pad drivers for even scrubbing on uneven floors.
  • Onboard battery charger for convenient recharging.
  • Green-certified equipment.