Learn More
Electron Microscopy Sciences Lab Grade Wide Mouth Lab Dewar, 350 mL with standard base, 86 mm O.D., 157mm height
SDP
Catalog No. 501930900
Encompass
These lab grade dewars are manufactured of borosilicate glass for liquid nitrogen service. A sturdy mesh cover facilitates a firm grip, provides a cushion against bumping and increases protection. Rated the most efficient glass dewars as a result of rigid quality control along with superior design and manufacturing features.
The weighted bases result in increased stability.
The dewars are supplied with a stopper to minimize evaporation
Evacuation tip is completely shielded for increased protection.
Efficiency is assured by rigid bakeout and evacuation to 10-5 torr or better. No pads between walls on the dewars eliminates heat transfer potential. Optional full length metal housing for added protection.
Volume: 350 mL
Type: Std Base
O.D.: 86 mm
Height: 157mm
Catalog No. 50-193-0900 Supplier Electron Microscopy Sciences Supplier No. 6188802
These lab grade dewars are manufactured of borosilicate glass for liquid nitrogen service. A sturdy mesh cover facilitates a firm grip, provides a cushion against bumping and increases protection. Rated the most efficient glass dewars as a result of rigid quality control along with superior design and manufacturing features.
The weighted bases result in increased stability.
The dewars are supplied with a stopper to minimize evaporation
Evacuation tip is completely shielded for increased protection.
Efficiency is assured by rigid bakeout and evacuation to 10-5 torr or better. No pads between walls on the dewars eliminates heat transfer potential. Optional full length metal housing for added protection.
Volume: 350 mL
Type: Std Base
O.D.: 86 mm
Height: 157mm