Learn More
TARGETMOL CHEMICALS INC CYM-5541 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T396125MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CYM-5541 (ML249) is a selective and allosteric S1P3 receptor agonist. purity: 98%
Catalog No. 50-258-0045