Learn More
Medchemexpress LLC HY-101419 5mg Medchemexpress, CYM-5541 CAS:945128-26-7 Purity:>98%
SDP
Catalog No. 501932119
Encompass_Preferred
Medchemexpress, HY-101419 5mg CYM-5541 CAS:945128-26-7 CYM-5541 (ML249) is an selective and allosteric S1P 3 receptor agonist with an EC 50 between 72 and 132 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2119 Supplier Medchemexpress LLC Supplier No. HY1014195MG
Medchemexpress, HY-101419 5mg CYM-5541 CAS:945128-26-7 CYM-5541 (ML249) is an selective and allosteric S1P 3 receptor agonist with an EC 50 between 72 and 132 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.