Learn More
Medchemexpress LLC HY-107735 5mg Medchemexpress, CYM 9484 CAS:1383478-94-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1077355MG
Encompass_Preferred
Medchemexpress, HY-107735 5mg CYM 9484 CAS:1383478-94-1 CYM 9484 is a selective and highly potent neuropeptide Y (NPY) Y2 receptor antagonist with an IC50 value of 19 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8460