Learn More
Medchemexpress LLC HY-108494 5mg Medchemexpress, CYM50260 CAS:1355026-60-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084945MG
Encompass_Preferred
Medchemexpress, HY-108494 5mg CYM50260 CAS:1355026-60-6 CYM50260 is a potent and exquisitely selective sphingosine-1-phosphate 4 receptor (S1P4-R) agonist with an EC50 of 45 nM. CYM50260 displays no activity against S1P1-R, S1P2-R, S1P3-R and S1P5-R. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8559