Learn More
Medchemexpress LLC CYM50358, Solution, 10 mM * 1 mL in DMSO
SDP
Catalog No. NC3245637
Encompass_Preferred
CYM50358 is a potent and selective S1PR4 antagonist, with an IC50 of 25 nM. CYM50358 can be used for the research of influenza infection.
Catalog No. NC3245637 Supplier Medchemexpress LLC Supplier No. HY13646210MM1ML DMSO
CYM50358 is a potent and selective S1PR4 antagonist, with an IC50 of 25 nM. CYM50358 can be used for the research of influenza infection.