Learn More
Zymo Research Corporation ZymoPURE II™ Plasmid Gigaprep (5 preps)
SDP
Catalog No. NC0820534
Encompass_Preferred
The ZymoPURE™ technology uses a modified alkaline lysis method and features novel binding chemistry, which enables the highest yields and concentration of plasmidDNA (up to 3 g/l) directly from a spin-column. Coupling ZymoPURE with the innovative EndoZero™ Spin-Columns, to eliminate endotoxins, achieves EndoZeroplasmid DNA ( 0.025 EU/g of plasmid DNA), making it suitable for transfection, restriction endonuclease digestion, in vivo studies, bacterial transformation, PCRamplification, DNA sequencing, and other sensitive downstream applications.As an added convenience, the ZymoPURE™ II Plasmid Gigaprep Kit contains colored buffers that permit error-free visualization and identification of completebacterial cell lysis and neutralization. Also, bottle top filters are included for rapid clearing of the lysate.
Catalog No. NC0820534 Supplier Zymo Research Corporation Supplier No. D4204
The ZymoPURE™ technology uses a modified alkaline lysis method and features novel binding chemistry, which enables the highest yields and concentration of plasmidDNA (up to 3 g/l) directly from a spin-column. Coupling ZymoPURE with the innovative EndoZero™ Spin-Columns, to eliminate endotoxins, achieves EndoZeroplasmid DNA ( 0.025 EU/g of plasmid DNA), making it suitable for transfection, restriction endonuclease digestion, in vivo studies, bacterial transformation, PCRamplification, DNA sequencing, and other sensitive downstream applications.As an added convenience, the ZymoPURE™ II Plasmid Gigaprep Kit contains colored buffers that permit error-free visualization and identification of completebacterial cell lysis and neutralization. Also, bottle top filters are included for rapid clearing of the lysate.