Learn More
Apexbio Technology LLC CYMOXANIL-500MG
SDP
Catalog No. 5000568813
Encompass_Preferred
Cymoxanil (CAS No 57966-95-7) is a fungicide primarily used within agricultural research Originating from a class of cyanoacetamide oximes its mechanism of action involves the inhibition of fungal growth by disrupting the spore germination process and interfering with the normal function of fungal mitochondrial pathways Its in vitro activity is commonly observed within the nanomolar to low micromolar range demonstrating efficacy in various biological assays Cymoxanil is typically applied in studies involving plant tissue cultures greenhouse trials and field studies where it serves as a crucial component of integrated pest management systems Additionally it is often utilized in drug screening programs to evaluate antifungal potentials Experimental concentrations are typically determined based on the specific research design with consideration for factors such as pathogen type environmental conditions and target crop species
Catalog No. 50-005-68813 Supplier Apexbio Technology LLC Supplier No. C6555500
Cymoxanil (CAS No 57966-95-7) is a fungicide primarily used within agricultural research Originating from a class of cyanoacetamide oximes its mechanism of action involves the inhibition of fungal growth by disrupting the spore germination process and interfering with the normal function of fungal mitochondrial pathways Its in vitro activity is commonly observed within the nanomolar to low micromolar range demonstrating efficacy in various biological assays Cymoxanil is typically applied in studies involving plant tissue cultures greenhouse trials and field studies where it serves as a crucial component of integrated pest management systems Additionally it is often utilized in drug screening programs to evaluate antifungal potentials Experimental concentrations are typically determined based on the specific research design with consideration for factors such as pathogen type environmental conditions and target crop species