Learn More
Medchemexpress LLC Cymoxanil | 57966-95-7 | 99.6% | 5 G
SDP
Catalog No. 5000411612
Encompass_Preferred
Cymoxanil is a fungicidal cyanooxime that targets plant diseases caused by fungi belonging to the Perenosporales. It works by affecting the growth, DNA, and RNA synthesis in Phytophthora.
  • Fungicidal cyanooxime.
  • Targets plant diseases caused by fungi belonging to the Perenosporales.
  • Affects growth, DNA, and RNA synthesis in Phytophthora.
Catalog No. 50-004-11612 Supplier Medchemexpress LLC Supplier No. HYB20675G
Cymoxanil is a fungicidal cyanooxime that targets plant diseases caused by fungi belonging to the Perenosporales. It works by affecting the growth, DNA, and RNA synthesis in Phytophthora.
  • Fungicidal cyanooxime.
  • Targets plant diseases caused by fungi belonging to the Perenosporales.
  • Affects growth, DNA, and RNA synthesis in Phytophthora.