Learn More
Medchemexpress LLC Cymoxanil | 57966-95-7 | MFCD00137381 | 99.4% | 198.18 g/mol | C7H10N4O3 | 10 MG
SDP
Catalog No. 5000297477
Encompass_Preferred
Cymoxanil standard is an analytical reference material of the fungicide cymoxanil supplied as a small vial for laboratory use. It is formulated for chromatographic and mass spectrometric method development, calibration, and quality control in pesticide residue and environmental analysis.
  • Analytical standard suitable for HPLC and LC-MS.
  • High HPLC purity (≈99.4%) for reliable calibration.
  • Contains CAS number 57966-95-7 and molecular weight 198.18 g/mol.
  • Packaged as a 10 mg vial in brown glass for stable storage.
  • Batch-specific certificate of analysis (COA) provided with assay and purity data.
Catalog No. 50-002-97477 Supplier Medchemexpress LLC Supplier No. HYB2067R10MG
Cymoxanil standard is an analytical reference material of the fungicide cymoxanil supplied as a small vial for laboratory use. It is formulated for chromatographic and mass spectrometric method development, calibration, and quality control in pesticide residue and environmental analysis.
  • Analytical standard suitable for HPLC and LC-MS.
  • High HPLC purity (≈99.4%) for reliable calibration.
  • Contains CAS number 57966-95-7 and molecular weight 198.18 g/mol.
  • Packaged as a 10 mg vial in brown glass for stable storage.
  • Batch-specific certificate of analysis (COA) provided with assay and purity data.