Learn More
Medchemexpress LLC Cymoxanil | 57966-95-7 | MFCD00137381 | 99.6% | 500 MG
SDP
Catalog No. 5000410130
Encompass_Preferred
Cymoxanil is a fungicidal cyanooxime that targets plant diseases caused by fungi belonging to the Perenosporales. It works by impacting the growth, and DNA and RNA synthesis in Phytophthora.
  • Targets plant diseases caused by fungi belonging to the Perenosporales.
  • Impacts growth, DNA, and RNA synthesis in Phytophthora.
  • Highly sensitive to colony growth and germ tube emergence of Phytophthora infestans sporangia and encysted zoospores.
Catalog No. 50-004-10130 Supplier Medchemexpress LLC Supplier No. HYB2067500MG
Cymoxanil is a fungicidal cyanooxime that targets plant diseases caused by fungi belonging to the Perenosporales. It works by impacting the growth, and DNA and RNA synthesis in Phytophthora.
  • Targets plant diseases caused by fungi belonging to the Perenosporales.
  • Impacts growth, DNA, and RNA synthesis in Phytophthora.
  • Highly sensitive to colony growth and germ tube emergence of Phytophthora infestans sporangia and encysted zoospores.