Learn More
Medchemexpress LLC Cymoxanil (Standard) | 57966-95-7 | 99.4% | 50 MG
SDP
Catalog No. 5000411666
Encompass_Preferred
Cymoxanil (Standard) is the analytical standard of Cymoxanil, intended for research and analytical applications. This fungicidal cyanooxime targets plant diseases caused by fungi belonging to the Perenosporales, affecting growth, DNA, and RNA synthesis in Phytophthora.
  • Analytical standard
  • Intended for research and analytical applications
  • Fungicidal cyanooxime
  • Targets plant diseases caused by Perenosporales fungi
  • Affects growth, DNA and RNA synthesis in Phytophthora
Catalog No. 50-004-11666 Supplier Medchemexpress LLC Supplier No. HYB2067R50MG
Cymoxanil (Standard) is the analytical standard of Cymoxanil, intended for research and analytical applications. This fungicidal cyanooxime targets plant diseases caused by fungi belonging to the Perenosporales, affecting growth, DNA, and RNA synthesis in Phytophthora.
  • Analytical standard
  • Intended for research and analytical applications
  • Fungicidal cyanooxime
  • Targets plant diseases caused by Perenosporales fungi
  • Affects growth, DNA and RNA synthesis in Phytophthora