Learn More
TARGETMOL CHEMICALS INC CYN 154806 TFA 1MG
SDP
Catalog No. 502879421
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. CYN 154806 TFA is a cyclic octapeptide and an effective selective somatostatin sst2 receptor antagonist. For the recombinant sst2, sst1, sst3, sst4 and sst5 receptors, the pIC50 values are 8.58, 5.41, 6.07, 5.76 and 6.48, respectively. Purity 99.7%
Catalog No. 50-287-9421 Supplier TARGETMOL CHEMICALS INC Supplier No. TP13461MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. CYN 154806 TFA is a cyclic octapeptide and an effective selective somatostatin sst2 receptor antagonist. For the recombinant sst2, sst1, sst3, sst4 and sst5 receptors, the pIC50 values are 8.58, 5.41, 6.07, 5.76 and 6.48, respectively. Purity 99.7%