Learn More
Medchemexpress LLC HY-P1202A 1mg Medchemexpress, CYN 154806 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1202A1MG
Encompass_Preferred
Medchemexpress, HY-P1202A 1mg CYN 154806 (TFA) CAS: CYN 154806 (TFA), a cyclic octapeptide, is a potent and selective somatostatin sst2 receptor antagonist, with pIC 50 values of 8.58, 5.41, 6.07, 5.76 and 6.48 for human recombinant sst2, sst1, sst3, sst4 and sst5 receptors respectively [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4161