Learn More
Medchemexpress LLC CYN 154806 (TFA) | 2828432-46-6 | 98.3% | 1311.36 | 10 MG
SDP
Catalog No. 5000423490
Encompass_Preferred
CYN 154806 TFA is a cyclic octapeptide and a potent, selective somatostatin sst2 receptor antagonist.
  • Potent and selective somatostatin sst2 receptor antagonist
  • Exhibits pIC50 values for human recombinant sst2, sst1, sst3, sst4, and sst5 receptors
  • Inhibits SRIF-induced increases in extracellular acidification
  • Blocks SRIF-induced increases in [35S]-GTPγS binding
  • Reverses decreased acid response to CCh in M4 KO mice
Catalog No. 50-004-23490 Supplier Medchemexpress LLC Supplier No. HYP1202A10MG
CYN 154806 TFA is a cyclic octapeptide and a potent, selective somatostatin sst2 receptor antagonist.
  • Potent and selective somatostatin sst2 receptor antagonist
  • Exhibits pIC50 values for human recombinant sst2, sst1, sst3, sst4, and sst5 receptors
  • Inhibits SRIF-induced increases in extracellular acidification
  • Blocks SRIF-induced increases in [35S]-GTPγS binding
  • Reverses decreased acid response to CCh in M4 KO mice