Learn More
TARGETMOL CHEMICALS INC Cynandione A 1MG
SDP
Catalog No. 502877839
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cynandione A, an acetophenone derived from Cynanchum wilfordii, has anti-inflammatory activity and significantly reduces pro-inflammatory cytokine overexpression via macrophage alpha7 nAChR activation and subsequent IL-10 expression. Purity 99.81%
Catalog No. 50-287-7839 Supplier TARGETMOL CHEMICALS INC Supplier No. T702771MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cynandione A, an acetophenone derived from Cynanchum wilfordii, has anti-inflammatory activity and significantly reduces pro-inflammatory cytokine overexpression via macrophage alpha7 nAChR activation and subsequent IL-10 expression. Purity 99.81%