Learn More
TARGETMOL CHEMICALS INC CYNAROSIDE 10MG
SDP
Catalog No. 502576955
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cynaroside (Luteolin 7-O-Glucoside) is a flavone a flavonoid-like chemical compound. It has an anti-oxidant effect inhibiting lipid and protein oxidation. purity: 99%
Catalog No. 50-257-6955 Supplier TARGETMOL CHEMICALS INC Supplier No. T337610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cynaroside (Luteolin 7-O-Glucoside) is a flavone a flavonoid-like chemical compound. It has an anti-oxidant effect inhibiting lipid and protein oxidation. purity: 99%