Learn More
Cayman Chemical Cyanidin 3-O-β-D-Galactopyranoside (chloride), Purity: ≥95%, CAS: 27661-36-5, 10 mg
SDP
Catalog No. 502964313
Encompass_Preferred
An anthocyanin flavonoid with antioxidant properties; scavenges radicals in DPPH, TEAC, ORAC, and hydrogen peroxide assays; decreases the release of LDH and the activation of caspase-3 in lymphocytes with t-butyl hydroperoxide-induced oxidative damage
Catalog No. 50-296-4313 Supplier Cayman Chemical Supplier No. 2226410 MG
An anthocyanin flavonoid with antioxidant properties; scavenges radicals in DPPH, TEAC, ORAC, and hydrogen peroxide assays; decreases the release of LDH and the activation of caspase-3 in lymphocytes with t-butyl hydroperoxide-induced oxidative damage