Learn More
Cayman Chemical Cyanidin (chloride), Purity: ≥98%, CAS: 528-58-5, 10 mg
SDP
Catalog No. 502969987
Encompass_Preferred
A natural anthocyanidin with significant antioxidant activity; has vasoprotective effects and can interfere with inflammation, carcinogenesis, obesity, and diabetes
Catalog No. 50-296-9987 Supplier Cayman Chemical Supplier No. 1480310 MG
A natural anthocyanidin with significant antioxidant activity; has vasoprotective effects and can interfere with inflammation, carcinogenesis, obesity, and diabetes