Learn More
Bon Opus Biosciences Recombinant Cynomolgus 5T4(C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C00S50UG
Encompass_Preferred
Recombinant Cynomolgus Trophoblast Glycoprotein is produced by our Mammalian expression system and the target gene encoding Thr35-Ser355 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q4R8Y9. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# C00S
Catalog No. 50-208-0321