Learn More
Bon Opus Biosciences Recombinant Cynomolgus Albumin (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103250UG
Encompass_Preferred
Recombinant Cynomolgus Albumin (C-His) is produced by Human Cells. The target gene encoding Asp25-Ala608 is expressed with a C-His tag. Accession# A2V9Z4. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1032
Catalog No. 50-217-2631