Learn More
iQ Biosciences Cynomolgus Monkey Bone Marrow Cells, 5M cells/vial
Catalog No. NC2855586
Encompass_Preferred
High-quality, high-viability (typically > 70%) cynomolgus (cyno) monkey (or macaque) bone marrow cells sourced responsibly from a vetted third party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor
Catalog No. NC2855586 Supplier iQ Biosciences Supplier No. IQBMNBM15
High-quality, high-viability (typically > 70%) cynomolgus (cyno) monkey (or macaque) bone marrow cells sourced responsibly from a vetted third party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor