Learn More
Bon Opus Biosciences Recombinant Cynomolgus CD16a (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW9150UG
Encompass_Preferred
Recombinant Cynomolgus Low Affinity Immunoglobulin Gamma Fc Region Receptor III-A is produced by our Mammalian expression system and the target gene encoding Glu21-Gly206 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# A0A140HDP8. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# CW91
Catalog No. 50-208-0468