Learn More
Bon Opus Biosciences Recombinant Cynomolgus B7-1/CD80 (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP17850UG
Encompass_Preferred
Recombinant Cynomolgus T-lymphocyte Activation Antigen CD80 is produced by our Mammalian expression system and the target gene encoding Val35-Asn242 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# G7NXN7. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, 5% Trehalose pH 7.4. Bon Opus Cat# BP178
Catalog No. 50-208-0283