Learn More
Bon Opus Biosciences Recombinant Cynomolgus B7-2/CD86 (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP18050UG
Encompass_Preferred
Recombinant Cynomolgus CD86 is produced by our Mammalian expression system and the target gene encoding Tyr25-His239 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# A0A2K5TTJ9. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,5% Trehalose, pH7.4. Bon Opus Cat# BP180
Catalog No. 50-208-0287