Learn More
Bon Opus Biosciences Recombinant Cynomolgus MELTF (N-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP107250UG
Encompass_Preferred
Recombinant Cynomolgus MELTF (N-hFc) is produced by Human Cells. The target gene encoding Gly111-Ala803 is expressed with a N-hFc tag. Accession# A0A2K5WMB7. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1072
Catalog No. 50-217-2670