Learn More
Bon Opus Biosciences Recombinan Cynomolgus MerTK (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP111150UG
Encompass_Preferred
Recombinan Cynomolgus MerTK (C-His) is produced by Human Cells. The target gene encoding Met1-Pro734 is expressed with a C-His tag. Accession# F6T7N0. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1111
Catalog No. 50-217-2707