Learn More
Bon Opus Biosciences Recombinant Cynomolgus MSLN (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP106650UG
Encompass_Preferred
Recombinant Cynomolgus MSLN (C-His) is produced by Human Cells. The target gene encoding Asp296-Ser598 is expressed with a C-His tag. Accession# A0A2K5TW94-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1066
Catalog No. 50-217-2664