Learn More
iQ Biosciences Cynomolgus Monkey CD20+ B Cells, 1M cells/vial
Catalog No. NC3591078
Encompass_Preferred
High-quality, high-viability (> 90%) cynomolgus (cyno) monkey (or macaque) CD20+ B Cells sourced responsibly from a vetted third party facility. Used in a variety of research areas including antibody production and evaluation of immune responses. All orders come with an iQ Certificate of Analysis.
Catalog No. NC3591078 Supplier iQ Biosciences Supplier No. IQBMN1B1
High-quality, high-viability (> 90%) cynomolgus (cyno) monkey (or macaque) CD20+ B Cells sourced responsibly from a vetted third party facility. Used in a variety of research areas including antibody production and evaluation of immune responses. All orders come with an iQ Certificate of Analysis.