Learn More
iQ Biosciences Cynomolgus Monkey Splenocytes, 25M cells/vial
Catalog No. NC1888137
Encompass_Preferred
High-quality, high-viability (typically > 70%) cynomolgus (cyno) monkey (or macaque) splenocytes sourced responsibly from a vetted third-party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor - Donor-matched PBMCs
Catalog No. NC1888137 Supplier iQ Biosciences Supplier No. IQBMNSP102
High-quality, high-viability (typically > 70%) cynomolgus (cyno) monkey (or macaque) splenocytes sourced responsibly from a vetted third-party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor - Donor-matched PBMCs