Learn More
Cayman Chemical Cyano-myracrylamide, Purity: ≥98%, CAS: 2801702-34-9, 10 mg
SDP
Catalog No. 502912538
Encompass_Preferred
A zDHHC20 inhibitor (IC50 = 1.35 µM in a cell-free assay); selectively inhibits zDHHC20 over APT-1 and APT-2 at 50 µM; inhibits S-acylation of Legionella E3 ligase GobX, MyD88, and Ras, which are substrates of zDHHC20, zDHHC9, and zDHHC6, respectively, in HEK293T cells expressing recombinant Legionella GobX, recombinant human MyD88, or endogenous Ras; is not toxic to HEK293T cells at 40 µM
Catalog No. 50-291-2538 Supplier Cayman Chemical Supplier No. 3515510 MG
A zDHHC20 inhibitor (IC50 = 1.35 µM in a cell-free assay); selectively inhibits zDHHC20 over APT-1 and APT-2 at 50 µM; inhibits S-acylation of Legionella E3 ligase GobX, MyD88, and Ras, which are substrates of zDHHC20, zDHHC9, and zDHHC6, respectively, in HEK293T cells expressing recombinant Legionella GobX, recombinant human MyD88, or endogenous Ras; is not toxic to HEK293T cells at 40 µM