Learn More
Bon Opus Biosciences Recombinant Cynomolgus ROR2 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP109450UG
Encompass_Preferred
Recombinant Cynomolgus ROR2 (C-hFc) is produced by Human Cells. The target gene encoding Val35-Gly403 is expressed with a C-hFc tag. Accession# A0A2K5UT30. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1094
Catalog No. 50-217-2692