Learn More
Bon Opus Biosciences Recombinant Cynomolgus Siglec-15 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW7050UG
Encompass_Preferred
Recombinant Cynomolgus Sialic Acid-binding Ig-like Lectin 15 is produced by our Mammalian expression system and the target gene encoding Phe20-Thr263 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# A0A2K5UY47. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,150mMNacl,0.3%chaps,5%Trehalose, pH7.4. Bon Opus Cat# CW70
Catalog No. 50-208-0451