Learn More
Bon Opus Biosciences Recombinant Cynomolgus OX40L/TNFSF4 (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP154500UG
Encompass_Preferred
Recombinant Cynomolgus OX40 Ligand is produced by our Mammalian expression system and the target gene encoding Gln51-Leu183 is expressed with a 8His tag at the N-terminus. Expression system: Human 293 Cells. Accession# F7FL80. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP154
Catalog No. 50-208-0244