Learn More
Bon Opus Biosciences Recombinant Cynomolgus TREM1 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103150UG
Encompass_Preferred
Recombinant Cynomolgus TREM1 (C-His) is produced by Human Cells. The target gene encoding Thr21-Pro201 is expressed with a C-His tag. Accession# A0A1D5RC96. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1031
Catalog No. 50-217-2630