Learn More
Sino Biological Cynomolgus ACVR2B Protein 200ug (Sino Biological)
Supplier:  Sino Biological 29662H27HB100
Encompass_Preferred
A DNA sequence encoding the cynomolgus ACVR2B (EHH16502.1) (Ser28-Thr143) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4460