Learn More
Sino Biological Cynomolgus ACVR2B Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90056CCCH100
Encompass_Preferred
A DNA sequence encoding the cynomolgus ACVR2B (EHH16502.1) (Ser28-Thr143) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-237-2136